Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Natural LDPE Boston Round Bottles Black with Phenolic PolyCone Lined Cap

Ideal solution for liquid sample handling in a squeezable bottle. Fisherbrand™ Natural LDPE Boston Round Bottles Black with Phenolic PolyCone Lined Cap are unbreakable and more resistant to stress cracking than HDPE bottles.

 View more versions of this product

Catalog No. 02-911-909A


  Check Availability
Add to cart

Includes: Phenolic PE Cone lined cap

Specifications

Description

  • Bottles retain their impact strength at very low temperatures
  • Caps form an tight seal and offer a good chemical barrier
  • Liner molds itself around the sealing areas of the finish as the cap is screwed on, virtually eliminating leakage, evaporation, contamination, binding and backing off
Description & Specifications

Specifications

480mL
Phenolic
LDPE
Bottle, Boston Round
PolyCone
28-400
Screw
16 oz.
Natural
24/Cs.
Black
Narrow
Round
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit