Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Natural Polypropylene Jars with White Polypropylene Unlined Cap Available on GSA/VA Contract for Federal Government customers only.

Ideal for storing and collecting specimens and lab supplies. Fisherbrand™ Natural Polypropylene Jars with White Polypropylene Unlined Cap are translucent and durable.

 View more versions of this product

Catalog No. 02-912-027A


  Check Availability
Add to cart

Includes: White Polypropylene unlined caps

Description

Description

Natural Polypropylene Jars are translucent and durable. Jars are ideal for storing and collecting specimens and lab supplies. Wide neck and straight sides facilitate easy recovery of contents. Polypropylene offers greater tolerance to high temperatures than HDPE and better contact clarity.

  • Features wide neck and straight sides to facilitate easy recovery of contents
  • Polypropylene material offers greater tolerance to high temperatures than HDPE and better contact clarity

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Specifications

960mL
Polypropylene
PolypropyleneþPolypropylene
Jar, Straight SidedþJar
Unlined
120-400
Screw
32 oz.
NaturalþNatural
24/Cs.
White
Wide
Straight Sided Jar
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit