Fisherbrand™ Natural Polypropylene Jars with White Polypropylene Unlined Cap
Ideal for storing and collecting specimens and lab supplies. Fisherbrand™ Natural Polypropylene Jars with White Polypropylene Unlined Cap are translucent and durable.
 View more versions of this product
Catalog No. 02-912-024A
  Check Availability
Add to Cart
Includes: Polypropylene unlined cap
    Specifications
    Capacity (Metric) 480mL
    Capacity (English) 16 oz.
    Product Type Straight Sided Jar
    Closure Color White
    Closure Material Polypropylene
    Closure Type Screw
    Color Natural
    Material Polypropylene
    Liner Material Unlined
    Mouth Wide
    Neck Finish 89-400
    Quantity 24/Cs.
    Shape Straight Sided Jar
    • Features wide neck and straight sides to facilitate easy recovery of contents
    • Polypropylene material offers greater tolerance to high temperatures than HDPE and better contact clarity
  • Description & Specifications
Related Products
Product Content Feedback