Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nav1.4 Na+ channel Mouse anti-Rat, Unconjugated, Clone: N255/38 , MilliporeSigma™

Mouse Monoclonal Antibody

$147.00 - $372.00

Specifications

Antigen Nav1.4 Na+ channel
Applications Immunohistochemistry (Paraffin)
Conjugate Unconjugated
Format Purified
Host Species Mouse
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MABN18511MI
View Documents
MilliporeSigma
MABN1851100UG
100 μg
Each for $372.00
Add to cart
 
MABN18512MI
View Documents
MilliporeSigma
MABN185125UG
25 μg
Each for $147.00
Add to cart
 
Description

Description

Nav1.4 Na+ channel Monoclonal antibody specifically detects Nav1.4 Na+ channel in Rat samples. It is validated for Immunohistochemistry (Paraffin).
Specifications

Specifications

Nav1.4 Na+ channel
Unconjugated
Mouse
Sodium channel protein type 4 subunit alpha;Mu-1;SkM1;Sodium channel protein skeletal muscle subunit alpha;Sodium channel protein type IV subunit alpha;Voltage-gated sodium channel subunit alpha Nav1.4
Recombinant fragment corresponding to 118 amino acids from the N-terminal half of Rat Voltage-gated sodium channel subunit alpha (Nav1.4).
Primary
Stable for 1 year at 2 to 8°C from date of receipt.
Immunohistochemistry (Paraffin)
Purified
Rat
Scn4a
Monoclonal
Protein G Purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit