Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nav1.6 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$138.37 - $320.71

Specifications

Antigen Nav1.6
Applications Western Blot
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB095475
View Documents
Novus Biologicals
NBP303464100UL
100 μg Each for $320.71
Add to cart
 
NB095476
View Documents
Novus Biologicals
NBP30346420UL
20 μg Each for $138.37
Add to cart
 
Description

Description

Nav1.6 Polyclonal antibody specifically detects Nav1.6 in Human samples. It is validated for Western Blot
Specifications

Specifications

Nav1.6
Unconjugated
Rabbit
Human
CerIII, hNa6/Scn8a voltage-gated sodium channel, MED, NaCh6, Nav1.6, PN4, sodium channel protein type 8 subunit alpha, Sodium channel protein type VIII subunit alpha, sodium channel, voltage gated, type VIII, alpha polypeptide, sodium channel, voltage gated, type VIII, alpha subunit, Voltage-gated sodium channel subunit alpha Nav1.6
Recombinant fusion protein containing a sequence corresponding to amino acids 1350-1440 of human Nav1.6 (NP_055006.1).
Polyclonal
Affinity purified
Western Blot
Purified
RUO
PBS with 50% glycerol, pH7.3.
6334
IgG
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit