Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NBAS Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA549534
Description
NBAS Polyclonal antibody specifically detects NBAS in Human samples. It is validated for Western Blot.Specifications
NBAS | |
Polyclonal | |
Unconjugated | |
NBAS | |
4933425L03Rik; ILFS2; NAG; NAG/BC035112 fusion; NAG/FAM49A fusion; NBAS; neuroblastoma amplified sequence; neuroblastoma-amplified gene protein; neuroblastoma-amplified protein; neuroblastoma-amplified protein-like; neuroblastoma-amplified sequence; similar to neuroblastoma-amplified protein; SOPH | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
51594 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide | |
A2RRP1 | |
NBAS | |
KLH conjugated synthetic peptide between 628-661 amino acids from the human NBAS. | |
200 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction