Learn More
Tocris Bioscience™ NBD 556
CD4 mimetic; blocks HIV-1 cell entry
$139.00 - $585.00
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
581110 View Documents Tocris Bioscience™
5811/10
Each for $139.00
Add to cart
581150 View Documents Tocris Bioscience™
5811/50
Each for $585.00
Add to cart
Description
Description
CD4 mimetic (IC50 values are in low micromolar range, dependent on cell type and HIV strain). Recognizes and induces structural changes in HIV-1 envelope protein gp120, analogous to CD4 binding. Inhibits HIV-1 cell entry in CXCR4 and CCR5 expressing cell lines, by blocking virus-cell and cell-cell fusion.