Learn More
Aat BioquestSupplier Diversity Partner NBD-X, succinimidyl ester *CAS 145195-58-0* DFS Item
Manufacturer:  Aat BioquestSupplier Diversity Partner 829
Compared to NBD chloride and fluoride, NBD-X acid gives better yield for labeling biopolymers. The fluorescence spectra of NBD derivatives are highly environment-sensitive, the fluorescence intensity decreases significantly in aqueous solutions.
Catalog No. 50-195-2955