Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ NBI 31772
High affinity insulin-like growth factor-I binding protein IGFBP inhibitor
$316.68 - $1280.64
Specifications
Purity | >98% |
---|---|
Description | NBI 31772 |
Molecular Weight (g/mol) | 341.27 |
Product Type | NBI 31772 |
Description
High affinity insulin-like growth factor-1 binding protein (IGFBP) inhibitor (Ki values are 1 - 24 nM for all six human subtypes). Displaces insulin-like growth factor-1 (IGF-1) from the IGF-1:IGFBP complex and suppresses IGF1-induced proliferation of 3T3 fibroblasts. Also increases cardiomyocyte proliferation in vivo.Specifications
>98% | |
NBI 31772 | |
341.27 | |
NBI 31772 |
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title