Learn More
NBL1 Rabbit anti-Human, Polyclonal, Bioss Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  BiossSupplier Diversity Partner BS6043R
Catalog No. 50-198-6999
Add to cart
Description
Description
This gene product is the founding member of the evolutionarily conserved CAN family of proteins, which contain a domain resembling the CTCK motif found in a number of signaling molecules. These proteins are secreted, and act as BMP antagonists by binding to BMPs and preventing them from interacting with their receptors. They may thus play an important role during growth and development. Alternatively spliced transcript variants encoding distinct isoforms have been identified for this gene.
Specifications
Specifications
NBL1
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
Nbl1
Liquid
Rabbit
IgG
100 μL
-20°C
Primary
4681
Nbl1
1 μg/mL
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
P41271
D1S1733E; D4H1S1733E; Dan; DAN domain family member 1; Dana; DAND1; differential screening-selected gene aberrant in neuroblastoma; N03; NB; NBL1; neuroblastoma 1, DAN family BMP antagonist; neuroblastoma candidate region, suppression of tumorigenicity 1; Neuroblastoma suppressor of tumorigenicity 1; neuroblastoma, suppression of tumorigenicity 1; Neuroblastoma, suppression of tumorigenicity 1 (DNA segment human D1S1733E); NO3; Protein N03; Zinc finger protein DAN
KLH conjugated synthetic peptide derived from human NBL1/DAND1.
Protein A
RUO
Antibody
Polyclonal
Human