Learn More
NBN Mouse anti-Human, Clone: OTI3C12, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF801448
Catalog No. 50-166-5601
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The p95 gene (identical to NBS1 and nibrin) is a member of the hMre11/hRad50 double-strand break complex. This protein complex has been implicated in Nijmegen breakage syndrome, an autosomal recessive disorder marked by increased cancer incidence, cell cycle checkpoint deficits, and ionizing radiation sensitivity, thus revealing a direct molecular link between double-strand break repair and cell cycle checkpoint functions.
Specifications
Specifications
NBN
Monoclonal
Unconjugated
NBN
Lyophilized
NBN
Human recombit protein fragment corresponding to amino acids 183-460 of human NBN produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Immunohistochemistry (Paraffin), Western Blot
OTI3C12
PBS with 8% trehalose and no preservative; pH 7.3
O60934
AT-V1, AT-V2, ATV, NBS, NBS1, P95
Mouse
IgG2a
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
4683