Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
Influenza A H7N1 Mouse anti-Virus, Alexa Fluor 647, Clone: InA334, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP150816AF647
Catalog No. NBP150816AH
Description
Description
Influenza A H7N1 Monoclonal antibody specifically detects Antigen in Virus samples. It is validated for Enzyme Linked Immunosorbent Assay.
Specifications
Specifications
Influenza A H7N1
InA334
ELISA
Purified
Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with Influenza A H7N1 or with Influenza A/Netherlands/219/03 H7N7 virus derived from allantoic fluid of 10 days old embryonated eggs
Protein G purified
RUO
Primary
Virus
ELISA
Alexa Fluor 647
50mM Sodium Borate with 0.05% Sodium Azide
Mouse
IgG2a
0.25 ml
Store at 4C in the dark.
Monoclonal