Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Albumin Mouse, Alexa Fluor 405, Clone: HSA20, Novus Biologicals
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP179049AF405
Description
Albumin Monoclonal antibody specifically detects Antigen in Bovine (Negative), Human samples. It is validated for Enzyme Linked Immunosorbent Assay, Western Blotting.Specifications
Albumin | |
HSA20 | |
Western Blot, ELISA | |
Purified | |
ALB | |
Human serum albumin | |
Protein A purified | |
RUO | |
Store at 4C in the dark. | |
Monoclonal | |
No cross-reactivity with other human proteins tested. |
ELISA, Western Blot | |
Alexa Fluor 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
albumin, cell growth inhibiting protein 42, DKFZp779N1935, growth-inhibiting protein 20, PRO0883, PRO0903, PRO1341, serum albumin | |
Mouse | |
IgG1 | |
0.2 ml | |
Apoptosis, Cancer, Cellular Markers, Golgi Apparatus Markers, Stem Cell Markers | |
Primary | |
213 | |
Bovine (Negative), Human |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title