Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Albumin Antibody (HSA20), DyLight 755, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP179049IR
Description
Albumin Monoclonal specifically detects Albumin in Human, Bovine (Negative) samples. It is validated for Western Blot, ELISA.Specifications
Albumin | |
Monoclonal | |
DyLight 755 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
ALB | |
Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with Human serum albumin. | |
0.2 mL | |
Apoptosis, Cancer, Cellular Markers, Golgi Apparatus Markers, Stem Cell Markers | |
213 | |
Human, Bovine (Negative) | |
Purified |
Western Blot, ELISA | |
HSA20 | |
Western Blot, ELISA | |
albumin, cell growth inhibiting protein 42, DKFZp779N1935, growth-inhibiting protein 20, PRO0883, PRO0903, PRO1341, serum albumin | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
No cross-reactivity with other tested human proteins. Does not react with bovine serum albumin, egg white albumin or human alpha fetoprotein. | |
Store at 4C in the dark. | |
IgG1 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction