Learn More
p27/Kip1 Mouse, DyLight 680, Clone: DCS-72.F6 + KIP1/769, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP244497FR
Catalog No. NBP244497FR
Add to cart
Description
Description
p27/Kip1 Monoclonal antibody specifically detects Antigen in Human, Monkey, Mouse, Rat samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin), Western Blotting.
Specifications
Specifications
p27/Kip1
DCS-72.F6 + KIP1/769
Western Blot, Flow Cytometry, Immunohistochemistry-Paraffin
Purified
CDKN1B
Mouse recombinant p27 protein (DCS-72.F6); Recombinant human CDKN1B protein (KIP1/769)
Protein A or G purified
RUO
Store at 4C in the dark.
Monoclonal
Recognizes a 27kDa protein, identified as the p27Kip1, a cell cycle regulatory mitotic inhibitor. Its epitope spans between aa 83-204 of p27. It is highly specific and shows no cross-reaction with other related mitotic inhibitors. p27Kip1 functions as a negative regulator of G1 progression and has been proposed to function as a possible mediator of TGF- induced G1 arrest. p27Kip1 is a candidate tumor suppressor gene. This MAb co-precipitates cdk4 in complex p27Kip1 and is excellent for staining of formalin-fixed tissues.
Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin)
DyLight 680
50mM Sodium Borate with 0.05% Sodium Azide
CDKN4, cyclin-dependent kinase inhibitor 1B, cyclin-dependent kinase inhibitor 1B (p27, Kip1), Cyclin-dependent kinase inhibitor p27, KIP1P27KIP1, MEN1B, MEN4, p27Kip1
Mouse
IgG
0.1 ml
Breast Cancer, Cancer, Cell Cycle and Replication, DNA Repair, Prostate Cancer, Tumor Suppressors
Primary
1027
Human, Mouse, Rat, Monkey