Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ NBQX disodium salt

Potent AMPA antagonist; more water soluble form of NBQX (Cat. No. 0373)

$174.00 - $1495.24

Specifications

Chemical Name or Material 2,3-Dioxo-6-nitro-1,2,3,4-tetrahydrobenzo[f]quinoxaline-7-sulfonamide disodium salt
CAS 479347-86-9
Assay Percent Range >98%
Solubility Soluble to 100mM in water
Molecular Formula C12H6N4O6SNa2
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
10-441-0
View Documents
Tocris Bioscience™
1044/10
10 mg
Each for $356.12
Add to Cart
 
10-445-0
View Documents
Tocris Bioscience™
1044/50
50 mg
Each for $1,495.24
Add to Cart
 
10-441-R
View Documents
Tocris Bioscience™
1044/1
1 mg
Each for $174.00
Add to Cart
 
Description

Description

NBQX disodium salt is a selective and competitive AMPA and kainate receptor antagonist (IC50 = 0.15 μM and 4.8 μM, respectively). NBQX blocks the antidepressant effects of 8-Hydroxy-DPAT hydrobromide (Cat. No. 0529), decreases mTOR and BDNF levels. NBQX is neuroprotective, anticonvulsant, antinociceptive and active in vivo.

NBQX disodium salt is a more water-soluble form of NBQX (Cat. No. 0373).

Specifications

Specifications

2,3-Dioxo-6-nitro-1,2,3,4-tetrahydrobenzo[f]quinoxaline-7-sulfonamide disodium salt
>98%
C12H6N4O6SNa2
479347-86-9
Soluble to 100mM in water
Observed MW: 380.24
Safety and Handling

Safety and Handling

Recommended Storage : Store at -20°C

SDS
Documents

Documents

Product Certifications

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit