Learn More
Nbs1 Mouse anti-Human, Alexa Fluor™ 532, Clone: 7E4A2, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP226297AF532
Catalog No. NP226297A32
Add to cart
Description
Description
Nbs1 Monoclonal antibody specifically detects Nbs1 in Human samples. It is validated for Western Blot,Flow Cytometry,ELISA,Immunohistochemistry,Immunocytochemistry,Immunofluorescence.
Specifications
Specifications
Nbs1
7E4A2
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Knockout Validated
ATV, AT-V1, AT-V2, Cell cycle regulatory protein p95, FLJ10155, MGC87362, NBS, NBS1P95, nibrin, Nijmegen breakage syndrome 1 (nibrin), Nijmegen breakage syndrome protein 1, p95, p95 protein of the MRE11/RAD50 complex
Mouse
IgG2 Alpha
Breast Cancer, Cancer, Cell Cycle and Replication, Checkpoint signaling, Chromatin Research, DNA Double Strand Break Repair, DNA Repair, Homologous Recombination, Non homologous end joining, Tumor Suppressors
Primary
4683
ELISA, Flow Cytometry, Immunoassay, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot
Alexa Fluor™ 532
50 mM sodium borate with 0.05% sodium azide
NBN
Nbs1 Antibody (7E4A2) [Alexa Fluor 532] was made to partial recombinant human NBS1 (between residues 400-650) expressed in E. coli [Uniprot O60934]
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human