Learn More
NCAM-1/CD56 Mouse anti-Human, Alexa Fluor™ 350, Clone: 1006411, R&D Systems™
Mouse Monoclonal Antibody
Manufacturer:  R&D Systems FAB24084U100UG
Catalog No. FB24084U100
Add to cart
This item is not returnable. View return policy
Description
Description
NCAM-1/CD56 Monoclonal antibody specifically detects NCAM-1/CD56 in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
NCAM-1/CD56
1006411
Flow Cytometry 0.25-1 μg/106 cells
NP_001070150
CD56, CD56/ NCAM-1, CD56 antigen, MSK39, NCAM1, N-CAM-1, NCAM-1, NCAMantigen recognized by monoclonal 5.1H11, neural cell adhesion molecule 1, neural cell adhesion molecule, NCAM
Mouse myeloma cell line NS0-derived recombinant human NCAM-1/CD56, Leu20-Pro603 and Glu636-Asn741, Accession # NP_001070150
Protein A or G purified from hybridoma culture supernatant
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Monoclonal
Detects human NCAM-1/CD56 in direct ELISAs.
Flow Cytometry
Alexa Fluor 350
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Purified, Supernatant, Conjugated
Mouse
IgG1
100 μg
Primary
4684
Human