Learn More
NCAM-1/CD56 Rabbit anti-Human, Mouse, Rat, Alexa Fluor™ 750, Clone: 735, Novus Biologicals™
Rabbit Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP252669AF750
Catalog No. NP22669A70
Add to cart
Description
Description
NCAM-1/CD56 Monoclonal antibody specifically detects NCAM-1/CD56 in Human,Mouse,Rat samples. It is validated for Western Blot,Flow Cytometry,Immunohistochemistry.
Specifications
Specifications
NCAM-1/CD56
735
Western Blot, Flow Cytometry, Immunohistochemistry
CD56, CD56/ NCAM-1, CD56 antigen, MSK39, N-CAM-1, NCAM-1, NCAMantigen recognized by monoclonal 5.1H11, neural cell adhesion molecule 1, neural cell adhesion molecule, NCAM
Rabbit
IgG Kappa
Store at 4°C in the dark.
Monoclonal
Recognises Polysialic acid (PSA), a carbohydrate polymer attached to the neural cell adhesion molecule (NCAM), which shares molecular epitopes with homopolymers of (alpha)2,8-linked sialic acid of Escherichia coli K1.
Flow Cytometry, Immunohistochemistry, Western Blot
Alexa Fluor™ 750
50 mM sodium borate with 0.05% sodium azide
NCAM1
Homopolymers of alpha 2,8-linked sialic acid of Escherichia coli K1.
0.1 mL
Primary
4684
Human, Mouse, Rat