Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NCAM-L1 Rabbit anti-Rat, Mouse, Unconjugated, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
$192.35 - $452.71
Specifications
Antigen | NCAM-L1 |
---|---|
Concentration | Please refer to lot specific datasheet. |
Applications | Immunofluorescence, Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
ABN2181-25UG
|
MilliporeSigma
ABN218125UG |
25 μg |
Each for $192.35
|
|
|||||
ABN2181-100
|
MilliporeSigma
ABN2181100UG |
100 μg |
Each for $452.71
|
|
|||||
Description
Anti-NCAM-L1, Cat. No. ABN2181, is a highly specific rabbit polyclonal antibody that targets Neural cell adhesion molecule L1 and has been tested in Immunofluorescence, Immunoprecipitation, and Western Blotting.Specifications
NCAM-L1 | |
Immunofluorescence, Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
Neuroscience | |
Purified rabbit polyclonal antibody in PBS with 0.05% sodium azide. | |
NP_031723 | |
Purified full-length murine Neural cell adhesion molecule L1-like protein (NCAM-L1). | |
Primary | |
Stable for 1 year at 2-8°C from date of receipt. | |
Antibodies |
Please refer to lot specific datasheet. | |
Polyclonal | |
Purified | |
RUO | |
Rat, Mouse | |
Neural cell adhesion molecule L1-like protein;L1 adhesion molecule;Cell adhesion molecule with homology to L1CAM;Chl1-like protein;Close homolog of L1 | |
Chl1;Call | |
IgG | |
Protein A purified | |
This rabbit polyclonal antibody detects NCAM-L1 protein in mouse and rat. | |
∽220 kDa observed; 135.07 kDa calculated. Uncharacterized bands may be observed in some lysate(s). |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title