Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NCAM1 Mouse anti-Human, Clone: NCAM1/795, Abnova™

Mouse monoclonal antibody raised against full length recombinant human NCAM1.

Manufacturer:  Abnova Corporation MAB13381

Catalog No. 89-934-658


Add to cart

Description

Description

Specifications

Specifications

NCAM1
NCAM1/795
Mouse monoclonal antibody raised against full length recombinant human NCAM1.
In 10mM PBS (0.05% BSA, 0.05% sodium azide).
CD56/MSK39/NCAM
Mouse
IgG1, kappa
100 ug
Store at 4°C.
Monoclonal
Human
Flow Cytometry, Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot
Unconjugated
Flow Cytometry (0.5-1 ug/106 cells in 0.1 mL) Immunofluorescence (1-2 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.5-1 ug/mL) Western Blot (0.5-1 ug/mL) The optimal working dilution should be determ
Liquid
NCAM1
Recombinant protein corresponding to full length human NCAM1.
Protein A/G purification
RUO
Primary
4684
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit