Learn More
NCAM1 Mouse anti-Human, Clone: NKI-nbl-1, Abnova™
Mouse monoclonal antibody raised against human NCAM1.
Manufacturer:  Abnova Corporation MAB12735
Catalog No. 89-934-660
Add to cart
Description
Description
Specifications
Specifications
NCAM1
NKI-nbl-1
Mouse monoclonal antibody raised against human NCAM1.
In PBS (0.09% sodium azide).
CD56/MSK39/NCAM
Mouse
IgG1
50 ug
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry (PFA fixed), Immunoprecipitation
Unconjugated
Immunofluorescence Immunohistochemistry (Frozen sections) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (20 ug/mL) Immunoprecipitation The optimal working dilution should be determined by the end user.
Liquid
NCAM1
Neuroblastoma cell line CHP-212
Protein G purification
RUO
Primary
4684