Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NCAM1 Rabbit anti-Human, Polyclonal , Abnova™

NCAM1 polyclonal antibody raised against recombinant human NCAM1.

Manufacturer:  Abnova Corporation PAB29492

Catalog No. 89-934-662


Add to cart

Description

Description

Sequence: DSENDFGNYNCTAVNRIGQESLEFILVQADTPSSPSIDQVEPYSSTAQVQFDEPEATGGVPILKYKAEWRAVGEEVWHSKWYDAK
Specifications

Specifications

NCAM1
Unconjugated
Immunofluorescence (1-4 ug/mL) Immunohistochemistry (1:50-1:200) The optimal working dilution should be determined by the end user.
Liquid
NCAM1
Recombinant protein corresponding to amino acids of human NCAM1.
Antigen affinity purification
RUO
Primary
4684
Immunofluorescence, Immunohistochemistry (PFA fixed)
NCAM1 polyclonal antibody raised against recombinant human NCAM1.
In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide)
CD56/MSK39/NCAM
Rabbit
IgG
100 uL
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
Human
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit