Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NCAPD2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA550367

Catalog No. PA550367


Add to cart

Description

Description

The antibody detects endogenous levels of total NCAPD2 protein.

NCAPD2 is the regulatory subunit of the condensin complex, a complex required for conversion of interphase chromatin into mitotic-like condense chromosomes. The condensin complex probably introduces positive supercoils into relaxed DNA in the presence of type I topoisomerases and converts nicked DNA into positive knotted forms in the presence of type II topoisomerases. NCAPD2 may target the condensin complex to DNA via its C-terminal domain.
Specifications

Specifications

NCAPD2
Polyclonal
Unconjugated
NCAPD2
Liquid
NCAPD2
Fusion protein corresponding to residues near the C terminal of human non-SMC condensin I complex, subunit D2
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunohistochemistry (Paraffin), Western Blot
1.7 mg/mL
PBS with 50% glycerol and 0.05% sodium azide; pH 7.3
Q15021
CAPD2, CNAP1, KIAA0159
Rabbit
IgG
100 μL
-20°C
Primary
9918
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit