Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NCAPD3 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA550368

Catalog No. PA550368


Add to cart

Description

Description

The antibody detects endogenous levels of total NCAPD3 protein.

This protein is a regulatory subunit of the condensin-2 complex, a complex which establishes mitotic chromosome architecture and is involved in physical rigidity of the chromatid axis.
Specifications

Specifications

NCAPD3
Polyclonal
Unconjugated
NCAPD3
Liquid
NCAPD3
Fusion protein corresponding to residues near the C terminal of human non-SMC condensin II complex, subunit D3
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunohistochemistry (Paraffin)
1.8 mg/mL
PBS with 50% glycerol and 0.05% sodium azide; pH 7.3
P42695
CAPD3, hcp-6, CAP-D3, hHCP-6, hCAP-D3
Rabbit
IgG
100 μL
-20°C
Primary
23310
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit