Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NCDN Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA597996
Description
NCDN Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA
Probably involved in signal transduction, in the nervous system, via increasing cell surface localization of GRM5 and positively regulating its signaling. Required for the spatial learning process. Acts as a negative regulator of Ca(2+)-calmodulin-dependent protein kinase 2 (CaMK2) phosphorylation. May play a role in modulating melanin-concentrating hormone-mediated functions via its interaction with MCHR1 that interferes with G protein-coupled signal transduction. May be involved in bone metabolism. May also be involved in neurite outgrowth.
Specifications
NCDN | |
Polyclonal | |
Unconjugated | |
NCDN | |
AU042419; Kiaa0607; minisatellite 10ae detected by probe MMS10; mKIAA0607; MMS10-AE; Ms10ae; M-Sema F-associating protein of 75 kDa; NCDN; Neurite outgrowth-related protein from the rat brain; neurochondrin; neurochondrin-1; neurochondrin-2; Norbin; Sfap75; similar to neurochondrin | |
Rabbit | |
Protein G | |
RUO | |
23154, 89791 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
2.57 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
O35095, Q9UBB6 | |
NCDN | |
Recombinant Human Neurochondrin protein (403-516AA). | |
100 μg | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction