Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NCE2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA531221
Description
Recommended positive controls: NT2D1. Predicted reactivity: Mouse (96%), Rat (83%), Zebrafish (81%), Xenopus laevis (88%), Chicken (92%), Rhesus Monkey (100%), Bovine (97%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Accepts the ubiquitin-like protein NEDD8 from the UBA3-NAE1 E1 complex and catalyzes its covalent attachment to other proteins. The specific interaction with the E3 ubiquitin ligase RBX2, but not RBX1, suggests that the RBX2-UBE2F complex neddylates specific target proteins, such as CUL5.
Specifications
NCE2 | |
Polyclonal | |
Unconjugated | |
UBE2F | |
2510010F15Rik; AI851109; MGC18120; NCE2; NEDD8 carrier protein UBE2F; NEDD8 protein ligase UBE2F; NEDD8-conjugating enzyme 2; NEDD8-conjugating enzyme UBE2F; RGD1307608; RING-type E3 NEDD8 transferase UBE2F; UBE2F; Ube2fa; ubiquitin conjugating enzyme E2 F (putative); ubiquitin conjugating enzyme E2F (putative); ubiquitin-conjugating enzyme E2 F; ubiquitin-conjugating enzyme E2F (putative) | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
140739 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
Q969M7 | |
UBE2F | |
Recombinant fragment corresponding to a region within amino acids 1 and 185 of Human NCE2. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction