Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NCF4 Mouse anti-Human, Clone: OTI12H10, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA527100

Catalog No. PIMA527100


Add to cart

Description

Description

Nicotinamide adenine dinucleotide phosphate (NADPH)-oxidase is a multimeric enzyme system that mediates electron transport from NADPH in the cytoplasm to molecular oxygen in the phagosome, thereby generating reactive oxidant intermediates. Upon neutrophil stimulation, NADPH-oxidase and other cytosolic elements localize to the cell membrane from the cytosol to form a complex which produces phagocytic oxygen radicals. There are a number of cytosolic proteins that are involved in NADPH-oxidase activation/ deactivation, including p47-phox, p67-phox, p40-phox and the small GTP-binding protein, Rac. Activation of NADPH oxidase is accompanied by the phosphorylation of cytosolic components p40-phox, p47-phox and p67-phox. The PKC consensus phosphorylation sights Thr 154 and Ser 315 in p40-phox are phosphorylated during activation of NADPH oxidase. p40-phox can promote oxidase activation by increasing the affinity of p47-phox for NADPH-oxidase. However, p40-phox appears to down-regulate oxidase function as well, by competing with an SH3 domain interaction between other essential oxidase components.
Specifications

Specifications

NCF4
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q15080
NCF4; P40PHOX; neutrophil cytosolic factor 4, 40kDa ; CTA-833B7.1; MGC3810; NCF; SH3PXD4; OTTHUMP00000028736; OTTHUMP00000043756; neutrophil NADPH oxidase factor 4; neutrophil cytosol factor 4; neutrophil cytosolic factor 4 (40kD)
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
4689
Immunohistochemistry (Paraffin)
OTI12H10
Unconjugated
NCF4
Liquid
NCF4
Full length human recombit protein of NCF4 produced in HEK293T cell
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit