Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NCOA1 Mouse anti-Human, Clone: 1135, Abnova™

Mouse monoclonal antibody raised against partial recombinant human NCOA1.

Manufacturer:  Abnova Corporation MAB12926

Catalog No. 89-934-668


Add to cart

Description

Description

The protein encoded by this gene acts as a transcriptional coactivator for steroid and nuclear hormone receptors. It is a member of the p160/steroid receptor coactivator (SRC) family and like other family members has histone acetyltransferase activity and contains a nuclear localization signal, as well as bHLH and PAS domains. The product of this gene binds nuclear receptors directly and stimulates the transcriptional activities in a hormone-dependent fashion. Alternatively spliced transcript variants encoding different isoforms have been identified. [provided by RefSeq]

Specifications

Specifications

NCOA1
1135
Mouse monoclonal antibody raised against partial recombinant human NCOA1.
In PBS, pH 7.4.
F-SRC-1/KAT13A/MGC129719/MGC129720/NCoA-1/RIP160/SRC-1/SRC1/bHLHe42/bHLHe74
Mouse
IgG1
50 uL
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
Immunohistochemistry (PFA fixed), Immunoprecipitation, Western Blot
Unconjugated
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (5 ug/mL) Immunoprecipitation Western Blot The optimal working dilution should be determined by the end user.
Liquid
NCOA1
Recombinant protein corresponding to amino acids 477-947 of human NCOA1.
Protein G purification
RUO
Primary
8648
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit