Learn More
NCOA3 Mouse anti-Human, Clone: AC3, Abnova™
Mouse monoclonal antibody raised against synthetic peptide of human NCOA3.
Manufacturer:  Abnova Corporation MAB12927
Catalog No. 89-934-669
Add to cart
Description
Description
The protein encoded by this gene is a nuclear receptor coactivator that interacts with nuclear hormone receptors to enhance their transcriptional activator functions. The encoded protein has histone acetyltransferase activity and recruits p300/CBP-associated factor and CREB binding protein as part of a multisubunit coactivation complex. This protein is initially found in the cytoplasm but is translocated into the nucleus upon phosphorylation. Two transcript variants encoding different isoforms have been found for this gene. In addition, a polymorphic repeat region is found in the C-terminus of the encoded protein. [provided by RefSeq]
Specifications
Specifications
NCOA3
AC3
Mouse monoclonal antibody raised against synthetic peptide of human NCOA3.
In PBS, pH 7.4.
ACTR/AIB-1/AIB1/CAGH16/CTG26/KAT13B/MGC141848/RAC3/SRC3/TNRC14/TNRC16/TRAM-1/bHLHe42/pCIP
Mouse
IgG1
50 uL
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
Immunohistochemistry (PFA fixed), Western Blot
Unconjugated
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (10 ug/mL) Western Blot The optimal working dilution should be determined by the end user.
Liquid
NCOA3
A synthetic peptide (conjugated with KLH) corresponding to C-terminus region of human NCOA3.
Protein G purification
RUO
Primary
8202