Learn More
NCOA3 Rabbit anti-Human, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 200321AP
Catalog No. 50-173-2664
Add to cart
Description
Description
The antibody can recognizes all the isoforms of NCOA3.
AIB1-AC3 recognizes AIB1, a member of the SRC-1 family of nuclear receptor coactivators with an apparent molecular weight of 160kDa. The AIB1 gene has been mapped to region 20q12 and has been shown to be amplified in breast tumors and pancreatic cancer cell lines. AIB1 interacts in a ligand-dependent fashion with the estrogen receptor to enhance estrogen-dependent transcriptional gene activation.
Specifications
Specifications
NCOA3
0.27 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9Y6Q9
ACTR, AIB 1, AIB1, CBP interacting protein, KAT13B, NCoA 3, NCOA3, nuclear receptor coactivator 3, pCIP, SRC 3, SRC3, TRAM 1, TRAM1
Rabbit
IgG
150 μL
-20°C
Primary
8202
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
NCOA3
Liquid
NCOA3
Peptide
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human