Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NCOA4 Mouse anti-Human, DyLight™ 650, Clone: OTI2G4, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272895C
Description
NCOA4 Monoclonal specifically detects NCOA4 in Human samples. It is validated for Western Blot.Specifications
NCOA4 | |
Monoclonal | |
DyLight 650 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
NCOA4 | |
Recombinant protein fragment corresponding to amino acids 280-553 of human NCOA4 (NP_001138732) produced in E.coli. | |
0.1 mL | |
8031.0 | |
Store at 4°C in the dark. |
Western Blot | |
OTI2G4 | |
Western Blot | |
Androgen receptor coactivator 70 kDa protein, Androgen receptor-associated protein of 70 kDa, ARA70RFGret fused, DKFZp762E1112, ELE1NCoA-4, nuclear receptor coactivator 4,70 kDa AR-activator, PTC3RET-activating gene ELE1, Ret-activating protein ELE1,70 kDa androgen receptor coactivator | |
Mouse | |
Affinity Purified | |
Primary | |
Human | |
IgG1 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title