Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NCOA7 Mouse anti-Human, Clone: OTI2F4, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA527307

Catalog No. PIMA527307


Add to cart

Description

Description

NCOA7 enhances the transcriptional activities of several nuclear receptors. It is involved in the coactivation of different nuclear receptors, such as ESR1, THRB, PPARG and RARA.
Specifications

Specifications

NCOA7
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q8NI08
ERAP140, ESNA1, Nbla00052, Nbla10993, dJ187J11.3, 140 kDa estrogen receptor-associated protein, estrogen nuclear receptor coactivator 1, estrogen receptor associated protein 140 kDa, putative protein product of Nbla00052, putative protein product of Nbla10993
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
135112
Western Blot
OTI2F4
Unconjugated
NCOA7
Liquid
NCOA7
Full length human recombit protein of NCOA7 produced in HEK293T cell
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit