Learn More
NCoR Rabbit anti-Human, Mouse, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A301145A
Description
The recommended shelf life for this product is 1 year from date of receipt.
This gene encodes a protein that mediates ligand-independent transcription repression of thyroid-hormone and retinoic-acid receptors by promoting chromatin condensation and preventing access of the transcription machinery. It is part of a complex which also includes histone deacetylases and transcriptional regulators similar to the yeast protein Sin3p. This gene is located between the Charcot-Marie-Tooth and Smith-Magenis syndrome critical regions on chromosome 17. Alternate splicing results in multiple transcript variants. Pseudogenes of this gene are found on chromosomes 17 and 20.Specifications
| NCoR | |
| Polyclonal | |
| Unconjugated | |
| NCOR1 | |
| Nuclear Receptor Corepressor, NCOR1, TRAC1, KIAA1047, hCIT529I10 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 20185, 9611 | |
| 4° C | |
| Liquid |
| ChIP sequencing (ChIP-seq), Immunoprecipitation, Western Blot | |
| 0.2 mg/ml | |
| TBS with 0.1% BSA and 0.09% sodium azide | |
| O75376, Q60974 | |
| NCOR1 | |
| Between 1525 and 1575 | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.