Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NCS-382, Tocris Bioscience™

Click to view available options
Quantity:
10mg
50mg
520505-01-5
C13H14O3
218.252
UADPGHINQMWEAG-FROQITRMSA-N
tocris-0780,2e-2-5s-5-hydroxy-5,7,8,9-tetrahydrobenzo 7 annulen-6-ylidene acetic acid,s,e-2-5-hydroxy-8,9-dihydro-5h-benzo 7 annulen-6 7h-ylidene acetic acid
6604757
(2E)-2-[(5S)-5-hydroxy-5,7,8,9-tetrahydrobenzo[7]annulen-6-ylidene]acetic acid
C1CC2=CC=CC=C2C(C(=CC(=O)O)C1)O
520505-01-5
C13H14O3
218.252
UADPGHINQMWEAG-FROQITRMSA-N
tocris-0780,2e-2-5s-5-hydroxy-5,7,8,9-tetrahydrobenzo 7 annulen-6-ylidene acetic acid,s,e-2-5-hydroxy-8,9-dihydro-5h-benzo 7 annulen-6 7h-ylidene acetic acid
6604757
(2E)-2-[(5S)-5-hydroxy-5,7,8,9-tetrahydrobenzo[7]annulen-6-ylidene]acetic acid
C1CC2=CC=CC=C2C(C(=CC(=O)O)C1)O

Antagonist of γ-hydroxybutyric acid

γ-Hydroxybutyric acid antagonist, anticonvulsant.
Quantity 50mg
Formula Weight 218.25
Percent Purity >99%
Chemical Name or Material NCS-382

Recommended Storage : Desiccate at +4°C