Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NCS-382, Tocris Bioscience™ Available on GSA/VA Contract for Federal Government customers only.

Click to view available options
Quantity:
10mg
50mg
520505-01-5
C13H14O3
218.252
UADPGHINQMWEAG-FROQITRMSA-N
tocris-0780,2e-2-5s-5-hydroxy-5,7,8,9-tetrahydrobenzo 7 annulen-6-ylidene acetic acid,s,e-2-5-hydroxy-8,9-dihydro-5h-benzo 7 annulen-6 7h-ylidene acetic acid
6604757
(2E)-2-[(5S)-5-hydroxy-5,7,8,9-tetrahydrobenzo[7]annulen-6-ylidene]acetic acid
C1CC2=CC=CC=C2C(C(=CC(=O)O)C1)O
520505-01-5
C13H14O3
218.252
UADPGHINQMWEAG-FROQITRMSA-N
tocris-0780,2e-2-5s-5-hydroxy-5,7,8,9-tetrahydrobenzo 7 annulen-6-ylidene acetic acid,s,e-2-5-hydroxy-8,9-dihydro-5h-benzo 7 annulen-6 7h-ylidene acetic acid
6604757
(2E)-2-[(5S)-5-hydroxy-5,7,8,9-tetrahydrobenzo[7]annulen-6-ylidene]acetic acid
C1CC2=CC=CC=C2C(C(=CC(=O)O)C1)O

Antagonist of γ-hydroxybutyric acid

γ-Hydroxybutyric acid antagonist, anticonvulsant.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Quantity 10mg
Formula Weight 218.25
Percent Purity >99%
Chemical Name or Material NCS-382

Recommended Storage : Desiccate at +4°C