Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NCX 4040, Tocris Bioscience™

Click to view available options
Quantity:
10mg
50mg
This item is not returnable. View return policy
287118-97-2
C16H13NO7
331.28
CTHNKWFUDCMLIQ-UHFFFAOYSA-N
no-aspirin,2-acetyloxy benzoic acid 4-nitrooxy methyl phenyl ester,2-acetyloxy-4-nitrooxy methyl phenyl ester, benzoic acid,4-nitrooxy methyl phenyl 2-acetyloxy benzoate,4-nitrooxymethyl phenyl 2-acetoxybenzoate,ncx hplc,2-acetyloxy benzoic acid 4-nitroxymethyl phenyl ester,2-acetyloxy benzoic acid 4-nitrooxymethyl phenylester
9818919
[4-(nitrooxymethyl)phenyl] 2-acetyloxybenzoate
CC(=O)OC1=CC=CC=C1C(=O)OC2=CC=C(C=C2)CO[N+](=O)[O-]
This item is not returnable. View return policy
287118-97-2
C16H13NO7
331.28
CTHNKWFUDCMLIQ-UHFFFAOYSA-N
no-aspirin,2-acetyloxy benzoic acid 4-nitrooxy methyl phenyl ester,2-acetyloxy-4-nitrooxy methyl phenyl ester, benzoic acid,4-nitrooxy methyl phenyl 2-acetyloxy benzoate,4-nitrooxymethyl phenyl 2-acetoxybenzoate,ncx hplc,2-acetyloxy benzoic acid 4-nitroxymethyl phenyl ester,2-acetyloxy benzoic acid 4-nitrooxymethyl phenylester
9818919
[4-(nitrooxymethyl)phenyl] 2-acetyloxybenzoate
CC(=O)OC1=CC=CC=C1C(=O)OC2=CC=C(C=C2)CO[N+](=O)[O-]

NO-donating aspirin; anti-inflammatory and antitumor

NO-donating aspirin; anti-inflammatory. Blocks inflammatory mediator production in isolated human monocytes (IC50 values are 0.07, 0.13, 0.15 and 0.18 μM for IL-1β, PGE2, IL-10 and TNF-α respectively). Decreases COX-2 expression (IC50 = 0.13 μM); disrupts proteasome-mediated degradation of iκB-α. Antiproliferative in HCT116 colon cancer cells; sensitizes drug-resistant ovarian tumor cells to cisplatin (Cat. No. 2251).
Quantity 10mg
Formula Weight 331.28
Percent Purity >98%
Chemical Name or Material NCX 4040

Recommended Storage : Store at -20°C