Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NCX 466, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
Description
Cyclooxygenase (COX)-inhibiting nitric oxide (NO) donor (CINOD). Inhibits COX-1 and COX-2 while releasing NO. Demonstrates higher efficacy than naproxen, its congener drug, in reducing levels of profibrotic cytokine transforming growth factor-β and oxidative stress in a mouse model of bleomycin-induced lung fibrosis. Orally available.
Specifications
Quantity | 10 mg |
Formula Weight | 436.41 |
Percent Purity | >98% |
Chemical Name or Material | NCX 466 |
Safety and Handling
Recommended Storage | Store at -20°C |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction