Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MT-ND3 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548356

Catalog No. PA548356


Add to cart

Description

Description

Core subunit of the mitochondrial membrane respiratory chain NADH dehydrogenase (Complex I) that is believed to belong to the minimal assembly required for catalysis. Complex I functions in the transfer of electrons from NADH to the respiratory chain. The immediate electron acceptor for the enzyme is believed to be ubiquinone.
Specifications

Specifications

MT-ND3
Polyclonal
PBS with 0.09% sodium azide
P03897
NADH-ubiquinone oxidoreductase chain 3, NADH dehydrogenase subunit 3, MT-ND3, MTND3, NADH3, ND3
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
4537
Flow Cytometry, Western Blot
Unconjugated
MT-ND3
Liquid
MT-ND3
KLH conjugated synthetic peptide between 10-38 amino acids from the N-terminal region of human ND3.
Antigen Affinity Chromatography, Protein A
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit