Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NDE1 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$436.00
Specifications
Antigen | NDE1 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP153097
|
Novus Biologicals
NBP153097 |
100 μL |
Each of 1 for $436.00
|
|
Description
NDE1 Polyclonal specifically detects NDE1 in Human samples. It is validated for Western Blot.Specifications
NDE1 | |
Polyclonal | |
Rabbit | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
FLJ20101, HOM-TES-87, nuclear distribution protein nudE homolog 1, NudE, nudE nuclear distribution gene E homolog 1 (A. nidulans), NUDE1, rat homolog | |
NDE1 | |
IgG | |
Affinity Purified |
Western Blot | |
Unconjugated | |
RUO | |
Q9NXR1-2 | |
54820 | |
Synthetic peptides corresponding to NDE1(nudE nuclear distribution gene E homolog 1 (A. nidulans)) The peptide sequence was selected from the middle region of NDE1. Peptide sequence AHRGPSSSLNTPGSFRRGLDDSTGGTPLTPAARISALNIVGDLLRKVGAL. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title