Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NDEL1 Mouse anti-Canine, Human, Rat, Clone: OTI1G10, liquid, TrueMAB™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  OriGeneSupplier Diversity Partner TA503058S

Catalog No. 50-167-4031


Add to cart

Description

Description

This gene encodes a thiol-activated peptidase that is phosphorylated in M phase of the cell cycle. Phosphorylation regulates the cell cycle-dependent distribution of this protein, with a fraction of the protein bound strongly to centrosomes in interphase and localized to mitotic spindles in early M phase. Overall, this protein plays a role in nervous system development. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.
Specifications

Specifications

NDEL1
Monoclonal
0.32 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q78PB6, Q9GZM8
EOPA, MITAP1, NDE1L1, NDE2, NUDEL
Mouse
IgG2b
30 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
170845, 489495, 81565
Flow Cytometry, Western Blot
OTI1G10
Unconjugated
NDEL1
Liquid
NDEL1
Full length human recombit protein of human NDEL1 produced in HEK293T cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Canine, Human, Rat
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit