Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Ndelta-Benzyloxycarbonyl-L-ornithine methyl ester hydrochloride, 98%

Click to view available options
Quantity:
1g
5g
25g
5874-75-9
C14H21ClN2O4
316.782
MFCD00077027
ZUJMKNWYEVTCNT-YDALLXLXSA-N
h-orn z-ome.hcl,h-orn z-ome hcl,h-orn z-ome-hcl,h-orn z-ome hydrochloride,c14h20n2o4.hcl,n-delta-z-l-ornithine methyl ester hydrochloride,h-orn cbz-ome inverted exclamation mark currencyhcl,methyl 2s-2-amino-5-benzyloxycarbonylaminopentanoate hydrochloride,l-ornithine,n5-phenylmethoxy carbonyl-, methyl ester, hydrochloride 1:1,methyl 2s-2-amino-5-benzyloxy carbonyl amino pentanoate hydrochloride
13967650
methyl (2S)-2-amino-5-(phenylmethoxycarbonylamino)pentanoate;hydrochloride
COC(=O)C(CCCNC(=O)OCC1=CC=CC=C1)N.Cl
5874-75-9
C14H21ClN2O4
316.782
MFCD00077027
ZUJMKNWYEVTCNT-YDALLXLXSA-N
h-orn z-ome.hcl,h-orn z-ome hcl,h-orn z-ome-hcl,h-orn z-ome hydrochloride,c14h20n2o4.hcl,n-delta-z-l-ornithine methyl ester hydrochloride,h-orn cbz-ome inverted exclamation mark currencyhcl,methyl 2s-2-amino-5-benzyloxycarbonylaminopentanoate hydrochloride,l-ornithine,n5-phenylmethoxy carbonyl-, methyl ester, hydrochloride 1:1,methyl 2s-2-amino-5-benzyloxy carbonyl amino pentanoate hydrochloride
13967650
methyl (2S)-2-amino-5-(phenylmethoxycarbonylamino)pentanoate;hydrochloride
COC(=O)C(CCCNC(=O)OCC1=CC=CC=C1)N.Cl
Beilstein 4341483
Formula Weight 316.78
Physical Form Solid
Percent Purity 98%
Melting Point 138°C to 141°C
Quantity 1g
Chemical Name or Material Ndelta-Benzyloxycarbonyl-L-ornithine methyl ester hydrochloride

storageNote1 : 2° to 8°C