Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Ndelta-Benzyloxycarbonyl-Nalpha-Fmoc-L-ornithine, 98%

Click to view available options
Quantity:
5g
25g
100g
138775-07-2
C28H28N2O6
488.54
MFCD00190893
QRBAKCWBDLHBLR-VWLOTQADSA-N
fmoc-orn z-oh,s-2-9h-fluoren-9-yl methoxy carbonyl amino-5-benzyloxy carbonyl amino pentanoic acid,n-alpha-9-fluorenylmethyloxycarbonyl-n-delta-benzyloxycarbonyl-l-ornithine,fmoc-orn z,ambotzfaa1731,nalpha-fmoc-ndelta-cbz-l-ornithine,n~5~-benzyloxy carbonyl-n~2~-9h-fluoren-9-yl methoxy carbonyl-l-ornithine,2s-5-benzyloxy carbonyl amino-2-9h-fluoren-9-ylmethoxy carbonyl amino pentanoic acid
14999635
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-5-(phenylmethoxycarbonylamino)pentanoic acid
C1=CC=C(C=C1)COC(=O)NCCCC(C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24
138775-07-2
C28H28N2O6
488.54
MFCD00190893
QRBAKCWBDLHBLR-VWLOTQADSA-N
fmoc-orn z-oh,s-2-9h-fluoren-9-yl methoxy carbonyl amino-5-benzyloxy carbonyl amino pentanoic acid,n-alpha-9-fluorenylmethyloxycarbonyl-n-delta-benzyloxycarbonyl-l-ornithine,fmoc-orn z,ambotzfaa1731,nalpha-fmoc-ndelta-cbz-l-ornithine,n~5~-benzyloxy carbonyl-n~2~-9h-fluoren-9-yl methoxy carbonyl-l-ornithine,2s-5-benzyloxy carbonyl amino-2-9h-fluoren-9-ylmethoxy carbonyl amino pentanoic acid
14999635
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-5-(phenylmethoxycarbonylamino)pentanoic acid
C1=CC=C(C=C1)COC(=O)NCCCC(C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24
Formula Weight 488.54
Physical Form Powder
Percent Purity 98%
Color White
Quantity 100g
Chemical Name or Material Ndelta-Benzyloxycarbonyl-Nalpha-Fmoc-L-ornithine

TSCA : No

storageNote1 : -30° to -10°C

Recommended Storage : Store at -20°C