Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NDFIP2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA552657
Description
Immunogen sequence: AAAGATGSEE LPPGDRGCRN GGGRGPAATT SSTGVAVGAE HGEDSLSRKP DPEPGRMDHH QPGTGRYQVL LNEEDN Highest antigen sequence identity to the following orthologs: Mouse - 68%, Rat - 74%.
Activates HECT domain-containing E3 ubiquitin-protein ligases, including ITCH, NEDD4, NEDD4L, SMURF2, WWP1 and WWP2, and consequently modulates the stability of their targets. As a result, may control many cellular processes. Recruits ITCH, NEDD4 and SMURF2 to endosomal membranes. Negatively regulates KCNH2 potassium channel activity by decreasing its cell-surface expression and interfering with channel maturation through recruitment of NEDD4L to the Golgi apparatus and multivesicular body where it mediates KCNH2 degradation (PubMed:26363003). May modulate EGFR signaling. Together with NDFIP1, limits the cytokine signaling and expansion of effector Th2 T-cells by promoting degradation of JAK1, probably by ITCH- and NEDD4L-mediated ubiquitination. [UniProt].
Specifications
NDFIP2 | |
Polyclonal | |
Unconjugated | |
NDFIP2 | |
0710001O20Rik; 2810436B12Rik; 9130207N19Rik; KIAA1165; MAPK-activating protein PM04 PM05 PM06 PM07; mKIAA1165; N4wbp5a; NDFIP2; Nedd4 family interacting protein 2; NEDD4 family-interacting protein 2; NEDD4 WW domain-binding protein 5A; NF-kappa-B-activating protein 413; putative MAPK-activating protein PM04/PM05/PM06/PM07; Putative NF-kappa-B-activating protein 413; Unknown (protein for IMAGE:7958216); wu:fa92h03; wu:fk31f07; zgc:63948 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
54602 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9NV92 | |
NDFIP2 | |
Recombinant protein corresponding to Human NDFIP2. Recombinant protein control fragment (Product #RP-89600). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction