Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NDM-1 Rabbit anti-Bacteria, E. coli, DyLight 350, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP177688UV

Catalog No. NBP177688UV


Add to cart

Description

Description

NDM-1 Polyclonal antibody specifically detects NDM-1 in Bacteria, E. coli samples. It is validated for Western Blot.
Specifications

Specifications

NDM-1
DyLight™ 350
50 mM sodium borate with 0.05% sodium azide
Rabbit
IgG
0.1 mL
Primary
Bacteria, E. coli
Western Blot
Western Blot
Affinity Purified, Conjugated
A synthetic peptide made to a C-terminal region of the E. coli bla-NDM-1 protein (within residues 100-158). [NCBI BAJ10873]
Immunogen affinity purified
Store at 4°C in the dark.
Polyclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit