Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NR1 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548855

Catalog No. PA548855

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Predicted to react with bovine based on sequence homology.

This gene encodes an NADPH-dependent diflavin reductase that contains both flavin mononucleotide (FMN) and flavin adenine dinucleotide (FAD) binding domains. The encoded protein is an enzyme that catalyzes the transfers electrons from NADPH through FAD and FMN cofactors to potential redox partners. Alternative splicing results in multiple transcript variants.
Specifications

Specifications

NR1
Polyclonal
PBS with 0.09% sodium azide
Q9UHB4, A2AI05
NADPH-dependent diflavin oxidoreductase 1, 16--, NADPH-dependent FMN and FAD-containing oxidoreductase, Novel reductase 1, NDOR1, NR1
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
27158, 78797
Western Blot
Unconjugated
NDOR1
Liquid
NDOR1
KLH conjugated synthetic peptide between 207-234 amino acids from the Central region of human NDOR1.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit