Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NDP52 Mouse anti-Human, FITC, Clone: 422, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP243696F

Catalog No. NBP243696F


Add to cart

Description

Description

NDP52 Monoclonal antibody specifically detects NDP52 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

NDP52
422
Immunocytochemistry/Immunofluorescence
Purified
CALCOCO2
Recombinant protein encompassing a sequence within the center region of human NDP52. The exact sequence is proprietary.
Protein G purified
Store at 4C in the dark.
Monoclonal
Human
Immunocytochemistry, Immunofluorescence
FITC
PBS with 0.05% Sodium Azide
Antigen nuclear dot 52 kDa protein, calcium binding and coiled-coil domain 2, calcium-binding and coiled-coil domain-containing protein 2, NDP52MGC17318, Nuclear domain 10 protein 52, Nuclear domain 10 protein NDP52, Nuclear dot protein 52
Mouse
IgG1
0.1 ml
Primary
10241
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit