Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NDRG1 Mouse anti-Human, Clone: NDRG1/1383, Abnova™

Mouse Monoclonal Antibody

Manufacturer:  Abnova Corporation MAB14721

Catalog No. 89-034-804

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Mouse monoclonal antibody raised against partial recombinant human NDRG1.

  • High specificity
  • High reactivity
Specifications

Specifications

NDRG1
NDRG1/1383
Mouse monoclonal antibody raised against partial recombinant human NDRG1.
In 10 mM PBS.
Liquid
NDRG1
Recombinant protein corresponding to amino acids 162-283 of human NDRG1.
Protein A/G purification
RUO
Primary
10397
Flow Cytometry, Immunofluorescence, Western Blot
Unconjugated
Flow Cytometry (0.5-1 ug/106 cells) Immunofluorescence (0.5-1 ug/mL) Western Blotting (0.5-1 ug/mL) The optimal working dilution should be determined by the end user.
Q92597
CAP43|CMT4D|DRG1|GC4|HMSNL|NDR1|NMSL|PROXY1|RIT42|RTP|TARG1|TDD5
Mouse
IgG3
100 ug
Store at -20 to -80°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only (RUO)