Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NDRG1 Mouse anti-Human, PE-Cyanine7, Clone: NDRG1/1383, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP254550PECY7

Catalog No. N25450PECY7


Add to cart

Description

Description

NDRG1 Monoclonal antibody specifically detects NDRG1 in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

NDRG1
NDRG1/1383
Flow Cytometry
Conjugated, Purified
NDRG1
Recombinant fragment of human NDRG1 fragment (aa162-283) (exact sequence is proprietary)
Protein A or G purified
Cancer, Phospho Specific
Primary
10397.0
Flow Cytometry
PE-Cyanine7
PBS with 0.05% Sodium Azide
CAP43, CMT4D, Differentiation-related gene 1 protein, DRG-1, DRG1HMSNL, NDR1, Nickel-specific induction protein Cap43, NMSL, N-myc downstream regulated 1, N-myc downstream-regulated gene 1 protein, protein NDRG1, protein regulated by oxygen-1, PROXY1, Reducing agents and tunicamycin-responsive protein, RIT42, RTPGC4, TARG1, TDD5, tunicamycin-responsive protein
Mouse
IgG3
0.1 mL
Store at 4°C in the dark. Do not freeze.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit