Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NDRG1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP30351620UL

 View more versions of this product

Catalog No. NB093056


Add to cart

Description

Description

NDRG1 Polyclonal antibody specifically detects NDRG1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunofluorescence, KnockDown
Specifications

Specifications

NDRG1
Unconjugated
Purified
Rabbit
IgG
20 μL
Cancer, Phospho Specific
Primary
10397
Immunofluorescence, KnockDown, Western Blot
PBS with 50% glycerol, pH7.3.
CAP43, CMT4D, Differentiation-related gene 1 protein, DRG-1, DRG1HMSNL, NDR1, Nickel-specific induction protein Cap43, NMSL, N-myc downstream regulated 1, N-myc downstream-regulated gene 1 protein, protein NDRG1, protein regulated by oxygen-1, PROXY1, Reducing agents and tunicamycin-responsive protein, RIT42, RTPGC4, TARG1, TDD5, tunicamycin-responsive protein
Recombinant fusion protein containing a sequence corresponding to amino acids 175-394 of human NDRG1 (NP_001128714.1).
Affinity purified
RUO
Store at -20°C. Avoid freeze-thaw cycles.
Polyclonal
Human, Mouse, Rat
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit